2-Bromoethylamine hydrobromide, 99%, Thermo Scientific Chemicals
2-Bromoethylamine hydrobromide, 99%, Thermo Scientific Chemicals
2-Bromoethylamine hydrobromide, 99%, Thermo Scientific Chemicals
2-Bromoethylamine hydrobromide, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Bromoethylamine hydrobromide, 99%, Thermo Scientific Chemicals
CAS: 2576-47-8 | C2H6BrN·HBr | 204.9 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
250 g
1 kg
Packaging:
Plastic bottle
Catalog number FSA106935000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS789-25-3
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
Appearance (Color)White
Assay (GC)≥96.0%
FormCrystals or powder or crystalline powder or lumps
Melting Point (clear melt)42.0-48.0?C