4-Bromoanisole, 98%, Thermo Scientific Chemicals
4-Bromoanisole, 98%, Thermo Scientific Chemicals
4-Bromoanisole, 98%, Thermo Scientific Chemicals
4-Bromoanisole, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Bromoanisole, 98%, Thermo Scientific Chemicals
CAS: 104-92-7 | C7H7BrO | 187.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
Catalog number FSA106631000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Request bulk or custom format
Chemical Identifiers
CAS331958-90-8
IUPAC Name4,4,5,5-tetramethyl-2-(oxolan-3-yl)-1,3,2-dioxaborolane
Molecular FormulaC10H19BO3
InChI KeyKEBDNHHRQNZXMA-UHFFFAOYNA-N
SMILESCC1(C)OB(OC1(C)C)C1CCOC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to light yellow
Appearance (Form)Liquid
GC>=96.0 %