Biuret, 97%, extra pure
Biuret, 97%, extra pure
Biuret, 97%, extra pure
Biuret, 97%, extra pure
Thermo Scientific Chemicals
Biuret, 97%, extra pure
CAS: 108-19-0 | C2H5N3O2 | 103.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Catalog number FSA106521000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Request bulk or custom format
Chemical Identifiers
CAS543-80-6
IUPAC Namebarium(2+) diacetate
Molecular FormulaC4H6BaO4
InChI KeyITHZDDVSAWDQPZ-UHFFFAOYSA-L
SMILES[Ba++].CC([O-])=O.CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Nitrate (NO3)=<20 ppm
Magnesium (Mg)=<10 ppm
Calcium (Ca)=<50 ppm
Appearance (Form)Crystalline powder
pH6.5 to 8.5 (5 % at 20°C)