2-Benzoylbenzoic Acid, 98+%, Thermo Scientific Chemicals
2-Benzoylbenzoic Acid, 98+%, Thermo Scientific Chemicals
2-Benzoylbenzoic Acid, 98+%, Thermo Scientific Chemicals
2-Benzoylbenzoic Acid, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Benzoylbenzoic Acid, 98+%, Thermo Scientific Chemicals
CAS: 85-52-9 | C14H10O3 | 226.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Plastic bottle
Catalog number FSA105710250
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS68818-86-0
IUPAC Name9,10-diethoxyanthracene
Molecular FormulaC18H18O2
InChI KeyGJNKQJAJXSUJBO-UHFFFAOYSA-N
SMILESCCOC1=C2C=CC=CC2=C(OCC)C2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream or yellow
FormCrystals or powder or crystalline powder or fused solid
Assay (HPLC)≥97.5%
Water Content (Karl Fischer Titration)≤2.5%
Melting Point (clear melt)141.0-153.0°C