Benzhydrol, 99%, Thermo Scientific Chemicals
Benzhydrol, 99%, Thermo Scientific Chemicals
Benzhydrol, 99%, Thermo Scientific Chemicals
Benzhydrol, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Benzhydrol, 99%, Thermo Scientific Chemicals
CAS: 91-01-0 | C13H12O | 184.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Plastic bottle
Catalog number FSA105395000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS21548-73-2
IUPAC Namedisilver(1+) sulfanediide
Molecular FormulaAg2S
InChI KeyXUARKZBEFFVFRG-UHFFFAOYSA-N
SMILES[S--].[Ag+].[Ag+]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Gray to black
FormPowder
Assay (special)Ag 86.8-87.0%
Total Metal Impurities≤0.005%