p-Anisidine, 99%, Thermo Scientific Chemicals
p-Anisidine, 99%, Thermo Scientific Chemicals
p-Anisidine, 99%, Thermo Scientific Chemicals
p-Anisidine, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
p-Anisidine, 99%, Thermo Scientific Chemicals
CAS: 104-94-9 | C7H9NO | 123.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
5 g
1 kg
Packaging:
Glass bottle
Catalog number FSA104832500
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS17639-76-8
IUPAC Namemethyl 2-methoxypropanoate
Molecular FormulaC5H10O3
InChI KeyVABBJJOSOCPYIT-UHFFFAOYNA-N
SMILESCOC(C)C(=O)OC
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
Refractive Index1.3945-1.3985 @ 20?C
Appearance (Color)Clear colorless to pale yellow