2-Aminopyrimidine, 98%, Thermo Scientific Chemicals
2-Aminopyrimidine, 98%, Thermo Scientific Chemicals
2-Aminopyrimidine, 98%, Thermo Scientific Chemicals
2-Aminopyrimidine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Aminopyrimidine, 98%, Thermo Scientific Chemicals
CAS: 109-12-6 | C4H5N3 | 95.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Plastic bottle
Catalog number FSA104581000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS62-56-6
IUPAC Namethiourea
Molecular FormulaCH4N2S
InChI KeyUMGDCJDMYOKAJW-UHFFFAOYSA-N
SMILESNC(N)=S
View more
SpecificationsSpecification SheetSpecification Sheet
Residue after ignition=<0.1 %
Solubility(16.5g in 100ml water,25°C) Passes test
Appearance (Color)White
Melting point174°C to 177°C
Titration Argentometric>=99.0 % (On dried basis)