4-Aminophenyl sulfone, 97%, Thermo Scientific Chemicals
4-Aminophenyl sulfone, 97%, Thermo Scientific Chemicals
4-Aminophenyl sulfone, 97%, Thermo Scientific Chemicals
4-Aminophenyl sulfone, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Aminophenyl sulfone, 97%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
100 g
Catalog number FSA104365000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Chemical Identifiers
CAS33454-82-9
IUPAC Nametrifluoromethanesulfonic acid lithium
Molecular FormulaCHF3LiO3S
InChI KeyWDGKXRCNMKPDSD-UHFFFAOYSA-N
SMILES[Li].OS(=O)(=O)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Lithium (Li)4.34 to 4.56 %