4-Aminophenyl ether, 98%, Thermo Scientific Chemicals
4-Aminophenyl ether, 98%, Thermo Scientific Chemicals
4-Aminophenyl ether, 98%, Thermo Scientific Chemicals
4-Aminophenyl ether, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Aminophenyl ether, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number FSA104330250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS5933-40-4
IUPAC Namemethyl(1-phenylethyl)amine
Molecular FormulaC9H13N
InChI KeyRCSSHZGQHHEHPZ-UHFFFAOYNA-N
SMILESCNC(C)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.5110 to 1.5130 (20°C, 589 nm)
Specific optical rotation+63° to +67° (20°C, 589 nm) (c=4, methanol)