2-Aminophenol, 99%, Thermo Scientific Chemicals
2-Aminophenol, 99%, Thermo Scientific Chemicals
2-Aminophenol, 99%, Thermo Scientific Chemicals
2-Aminophenol, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Aminophenol, 99%, Thermo Scientific Chemicals
CAS: 95-55-6 | C6H7NO | 109.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
Packaging:
Glass bottle
Catalog number FSA104261000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS951-55-3
IUPAC Name2-amino-3-(5-methyl-1H-indol-3-yl)propanoic acid
Molecular FormulaC12H14N2O2
InChI KeyHUNCSWANZMJLPM-UHFFFAOYSA-N
SMILESCC1=CC=C2NC=C(CC(N)C(O)=O)C2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Heavy metals (as Pb)=<10 ppm
Appearance of solution(5% soln. in 1 N NaOH) clear, pale yellowish
Infrared spectrumConforms
Water=<1 % (K.F.)