Allantoin, 98%, Thermo Scientific Chemicals
Allantoin, 98%, Thermo Scientific Chemicals
Allantoin, 98%, Thermo Scientific Chemicals
Allantoin, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Allantoin, 98%, Thermo Scientific Chemicals
CAS: 97-59-6 | C4H6N4O3 | 158.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number FSA102845000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Request bulk or custom format
Chemical Identifiers
CAS7727-21-1
IUPAC Namedipotassium [(sulfonatoperoxy)sulfonyl]oxidanide
Molecular FormulaK2O8S2
InChI KeyUSHAGKDGDHPEEY-UHFFFAOYSA-L
SMILES[K+].[K+].[O-]S(=O)(=O)OOS([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Titration Iodometric>=99.0 %
Chloride (Cl)=<120 ppm
Heavy metals (as Pb)=<50 ppm
Iron (Fe)=<20 ppm