9-Acetylanthracene, 95%, Thermo Scientific Chemicals
9-Acetylanthracene, 95%, Thermo Scientific Chemicals
9-Acetylanthracene, 95%, Thermo Scientific Chemicals
9-Acetylanthracene, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
9-Acetylanthracene, 95%, Thermo Scientific Chemicals
CAS: 784-04-3 | C16H12O | 220.27 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA102440050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS40397-98-6
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear to turbid colorless to yellow to gray
FormLiquid
Assay (GC)≥96.0%
Refractive Index1.5290-1.5340 @ 20?C