Check Order Status
Desiccator with Glass Knob
Actual product may vary
Desiccator with Glass Knob
Made from Simax 3.3 borosilicate glass. Fisherbrand Desiccator with Glass Knob is designed for variety of vacuum work.
Have Questions?
Catalog Number
Product Type
Diameter (Metric) Inner
Diameter (Metric) Outer
Height (Metric)
FKS15584635
Desiccator
239 mm
320 mm
363 mm
Catalog number
FKS15584635
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Product Type:
Desiccator
Diameter (Metric) Inner:
239 mm
Diameter (Metric) Outer:
320 mm
Height (Metric):
363 mm
Made from SIMAX 3.3 borosilicate glass
Designed for variety of vacuum work
Used for desiccation of samples, storage of materials sensitive to humidity
Specifications
Certifications/Compliance
ISO 13 130, DIN 12 491
Material
Simax Borosilicate Glass 3.3
Product Type
Desiccator
Includes
Glass knob
Height (Metric)
363 mm
Diameter (Metric) Inner
239 mm
Diameter (Metric) Outer
320 mm