Reusable Glass Media Bottles with Cap
Reusable Glass Media Bottles with Cap
Actual product may vary
Reusable Glass Media Bottles with Cap
Reusable Glass Media Bottles with Cap
Reusable Glass Media Bottles with Cap
Reusable Glass Media Bottles with Cap
Reusable Glass Media Bottles with Cap
Fisher Catalogue
Reusable Glass Media Bottles with Cap
Feature clear (natural), drip-free, replaceable polypropylene pour ring.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
FKS154161235000 mL
FKS15446113100 mL
FKS15456113250 mL
FISFB-800-500500 mL
FKS15476113500 mL
FISFB-800-10001000 mL
FISFB80010001000 mL
FKS154861131000 mL
FKS154061232000 mL
Catalog number FKS15416123
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
5000 mL
Use these glass media bottles for a variety of general laboratory applications. Fisherbrand™ Reusable Glass Media Bottles with Cap feature a convenient wide opening, enhanced graduations and a polypropylene pour ring. Feature clear (natural), drip-free, replaceable polypropylene pour ring.
  • Permanent white enamel marking spot
  • GL 45 screw thread
  • Linerless
  • Autoclavable (to 140°C)
  • Borosilicate glass
  • Polypropylene cap
Specifications
Capacity169.07 oz., 5000 mL
Capacity (Metric)5000 mL
Capacity (English)169.07 oz.
ColorNatural
Diameter (English)7.1 in.
Diameter (English) Inner Neck1.18 in.
Diameter (Metric) Inner Neck30 mm
IncludesClear drip-free Polypropylene pouring ring
MaterialGlass
No. per Case1
QuantityCase of 1
SterilityNon-sterile
Closure ColorBlue
Graduations1000 to 4000mL
Closure IncludedYes
AutoclavableAutoclavable
GraduatedYes
MouthWide
ShapeRound
Height (Metric)330 mm
Product TypeMedia Bottle
Height (English)12.9 in.
Closure MaterialPolypropylene
Closure SizeGL 45
Diameter (Metric)181 mm
Closure TypeScrew Cap
Unit Size