Check Order Status
Borosilicate Glass Tall Form Beakers
Actual product may vary
Fisher Catalogue
Borosilicate Glass Tall Form Beakers
Includes graduations and a spout for easy pouring. Fisherbrand™ Borosilicate Glass Tall Form Beakers are available in a variety of sizes.
Have Questions?
Change view
Catalog Number
Capacity (Metric)
FKS15409093
100 mL
FKS15419093
150 mL
FKS15429093
250 mL
FKS15449093
600 mL
4 Options
Catalog number
FKS15409093
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
100 mL
Borosilicate glass
Tall form
Graduated
With spout
Manufactured to ISO 3819, DIN 12331
Specifications
Color
Translucent
Diameter (Metric) Outer
48 mm
Capacity (Metric)
100 mL
Material
Borosilicate Glass
Finish
Matte
Height (Metric)
80 mm
Includes
Spout
Quantity
10 Pk.
Type
Beaker
Graduated
Yes
Certifications/Compliance
ISO 3819, DIN 12331
Scale
White Graduated Scale
Style
Tall Form
Unit Size