Search Thermo Fisher Scientific
Catalog Number | Diameter (Metric) | Quantity |
---|---|---|
FKS12494272 | 45 mm | 100/Pk. |
FKS11392805 | 90 mm | 100/Pk. |
FKS11302815 | 110 mm | 100/Pk. |
FKS10424081 | 125 mm | 100/Pk. |
FKS11322815 | 150 mm | 100/Pk. |
FKS10434271 | 185 mm | 100/Pk. |
FKS15788032 | 25 mm | 100/Pk. |
These cellulose filters are used in qualitative analytical techniques to determine and identify materials and for clarifying liquids.