Check Order Status
Aluminum Foil, Standard-Gauge Roll
Actual product may vary
Fisher Catalogue
Aluminum Foil, Standard-Gauge Roll
For sealing, weighing, and transferring samples
Have Questions?
Change view
Catalog Number
Length (English)
Thickness
Width (Metric)
Length (Metric)
FKS11759408
25 ft.
18 μm
30cm
7.6 m
FIS01-213-100
25 ft.
18 μm
30cm
7.6 m
2 Options
Catalog number
FKS11759408
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Length (English):
25 ft.
Thickness:
18 μm
Width (Metric):
30cm
Length (Metric):
7.6 m
Ideal for weighing, sealing, and tranferring your samples.
Packed in individual dispenser box with cutter bar
Heavy-gauge rolls offer greater folding strength, good for covering sharp edges
Specifications
Applications
Sealing, Weighing, Transferring Samples
Length (English)
25 ft.
Product Type
Foil
Width (English)
11.9 in.
Width (Metric)
30cm
Length (Metric)
7.6 m
Material
Aluminum
Thickness
18 μm
Unit Size