Stir Bar Box Set, Octahedral
Stir Bar Box Set, Octahedral
Actual product may vary
Fisherbrand
Stir Bar Box Set, Octahedral
For use in containers with bases that are slightly curved or uneven. Fisherbrand™ Star Bar Box Set, Octahedral is isostatically molded in pure, FDA-approved PTFE plastic.
Have Questions?
Catalog number FKS11547772
View Price:Sign in to see your account pricing. Need an account? Register with us today.
  • Guaranteed inert and leak-free
  • Individually polished to stay cleaner for longer
  • Each set contains:
2 each of 13 x 8mm
2 each of 15 x 8mm
2 each of 25 x 10mm
2 each of 38 x 10mm
2 each of 51 x 10mm
2 each of 64 x 10mm
Specifications
ColorWhite
Includes2 Followers Each: 113mm x 8mm, 16mm x 8mm, 16mm x 10mm, 25mm x 10mm, 38mm x 10mm, 51mm x 10mm, 64mm x 10mm
Unit Size