Sterilin™ Polystyrene Containers, 60mL to 250mL
Sterilin™ Polystyrene Containers, 60mL to 250mL
Actual product may vary
Fisherbrand
Sterilin™ Polystyrene Containers, 60mL to 250mL
Contain samples in Thermo Scientific™ Sterilin™ Polystyrene Containers with 60mL to 250mL capacities. Available with either plastic or metal flow seal caps.
Have Questions?
Catalog NumberCapacity (Metric)Closure MaterialLabelNo. per Case
FKS11389143150 mLMetalPrinted120/Cs.
Catalog number FKS11389143
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
150 mL
Closure Material:
Metal
Label:
Printed
No. per Case:
120/Cs.
Contain samples in Thermo Scientific™ Sterilin™ Polystyrene Containers with 60mL to 250mL capacities. Available with either plastic or metal flow seal caps.
NOT AVAILABLE IN AMERICA
Features:
  • Leak-tested in accordance with BS EN ISO 6717
Ordering Information:
Product is not available in North America.
Specifications
Product TypeContainer
Closure IncludedYes
MaterialPolystyrene
ShapeCylindrical
SterilityNon-sterile
Capacity (Metric)150 mL
Certifications/ComplianceLeak-tested in accordance with BS EN ISO 6717
Closure MaterialMetal
Diameter (Metric)55 mm
Height (Metric)108 mm
LabelPrinted
No. per Case120/Cs.
Unit Size