Pyrex™ Borosilicate Glass Baffled Erlenmeyer Flasks
Pyrex™ Borosilicate Glass Baffled Erlenmeyer Flasks
Actual product may vary
Fisher Catalogue
Pyrex™ Borosilicate Glass Baffled Erlenmeyer Flasks
Contains four indents to mix contents consistently and without risk of spillage. Pyrex™ Borosilicate Glass Baffled Erlenmeyer Flasks can withstand multiple autoclave cycles.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Height (Metric)Diameter (Metric) Outer
FKS113734541000 mL222 mm129 mm
FKS113834542000 mL282 mm161 mm
Catalog number FKS11373454
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
1000 mL
Height (Metric):
222 mm
Diameter (Metric) Outer:
129 mm
Contains four indents to mix contents consistently and without risk of spillage. Pyrex™ Borosilicate Glass Baffled Erlenmeyer Flasks can withstand multiple autoclave cycles.
  • Ideal for use in preparation of cultures
  • Manufactured from Pyrex™ Borosilicate Glass
  • Exhibits 4 baffle indents to provide optimal and consistent agitation
  • Suitable for repeated autoclave cycles at 121°C
Specifications
DescriptionConical; Style: Erlenmeyer
Capacity (Metric)1000 mL
MaterialBorosilicate Glass, Borosilicate Glass
For Use With (Application)Preparation of cultures
Height (Metric)222 mm
Neck StyleNarrow
ShapeConical
AutoclavableAutoclavable
Mouth StyleNarrow
Diameter (Metric) Outer129 mm
Unit Size