Azlon™ Polypropylene Graduated Molded Jugs with Handles
Azlon™ Polypropylene Graduated Molded Jugs with Handles
Actual product may vary
Fisher Catalogue
Azlon™ Polypropylene Graduated Molded Jugs with Handles
Manufactured in high clarity polypropylene. Azlon™ Polypropylene Graduated Moulded Jugs with Handles are tapered with ergonomically designed handle for comfort and control. Suitable for food applications in accordance with EU and US regulations. Permanent moulded graduations provide easy volume assessment. Autoclavable to 121° C.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Graduations
FKS11333694500 mL100mL
FKS113436941000 mL100mL
FKS113536942000 mL500mL
FKS113736945000 mL500mL
Catalog number FKS11333694
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
500 mL
Graduations:
100mL
Manufactured in high clarity polypropylene. Azlon™ Polypropylene Graduated Moulded Jugs with Handles are tapered with ergonomically designed handle for comfort and control. Suitable for food applications in accordance with EU and US regulations. Permanent moulded graduations provide easy volume assessment. Autoclavable to 121° C.
  • Tapered jugs in high clarity polypropylene
  • Permanent molded graduations
  • Autoclavable at 121° C
  • Ergonomically designed handle with thumb grip for easy carrying
Specifications
Capacity (Metric)500 mL
Capacity (English)16.91 oz.
ColorClear
MaterialPolypropylene
No. per PackIndividual
QuantityEach
Temperature Range (Metric)–20 to 135°C
Graduations100mL
Certifications/ComplianceEU; USA
Closure IncludedNo
AutoclavableAutoclavable
GraduatedYes
MouthWide
ShapeTapered
Height (Metric)140 mm
Product TypeMolded Jug
Height (English)5.52 in.
Diameter (Metric)90 mm
Through the Closure DispensingNo
Unit Size