Centrifugation Polypropylene Bio-Bottles
Centrifugation Polypropylene Bio-Bottles
Actual product may vary
Centrifugation Polypropylene Bio-Bottles
Use these polypropylene screw cap closure bio-bottles for compatibility with round-bucket centrifuge systems.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
FKS10482525400 mL
75003800
75007585
Catalog number FKS10482525
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
400 mL
Use these polypropylene screw cap closure bio-bottles for centrifugation compatibility with round buckets as in the Thermo Scientific™ TX-400 Rotor.
  • With screw cap
  • The 180mL Bio-Bottles (75-003-800) are compatible with Round Buckets for Thermo Scientific TX-200 Rotor (75-003-569)
  • The 400mL Bio-Bottles (75-007-585) are compatible with Round Buckets for Thermo Scientific TX-400 Rotor (75-003-655)
Specifications
Capacity12 x 400 mL
Capacity (Metric)400 mL
ColorClear
MaterialPolypropylene
Closure ColorBlue
AutoclavableAutoclavable
GraduatedNo
ShapeRound
Product TypeCentrifuge Bottle
For Use With (Equipment)Round Bucket for Thermo Scientific TX-400 Rotor
Closure MaterialPolypropylene
Closure TypeScrew Cap
Through the Closure DispensingNo
Unit SizePK