Check Order Status
Soda Lime Glass Short Stem Funnel
Actual product may vary
Fisher Catalogue
Soda Lime Glass Short Stem Funnel
Manufactured from soda lime glass. Fisherbrand™ Soda Lime Glass Short Stem Funnel is less likely to clog when funneling thicker substances.
Have Questions?
Change view
Catalog Number
Diameter (Metric)Top
Diameter (Metric) Stem
Length (Metric) Stem
FKS10123682
100mm
12mm
110mm
FKS10767414
80mm
9mm
85mm
2 Options
Catalog number
FKS10123682
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Diameter (Metric)Top:
100mm
Diameter (Metric) Stem:
12mm
Length (Metric) Stem:
110mm
Manufactured from Soda Lime glass.
Short stem funnel
Less likely to clog when funneling thicker substances.
Specifications
Diameter (Metric)Top
100mm
Diameter (Metric) Stem
12mm
Material
Soda Lime Glass
Height (Metric)
175 mm
For Use With (Application)
Ideal for filtering applications in the laboratory
Length (Metric) Stem
110mm
Unit Size