Nylon Test Tube Brush
Nylon Test Tube Brush
Actual product may vary
Nylon Test Tube Brush
Nylon Test Tube Brush
Nylon Test Tube Brush
Nylon Test Tube Brush
Nylon Test Tube Brush
With fan-shaped end.
Have Questions?
Catalog NumberColorLength (Metric)Diameter (Metric) Bristle
FISS13960White230 mm12 mm
Catalog number FISS13960
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Color:
White
Length (Metric):
230 mm
Diameter (Metric) Bristle:
12 mm
  • Made from nylon
  • Wire-wound handle with loop
Specifications
Product TypeTest Tube Brush
Bristle MaterialNylon
Bristle TypeSoft/Slightly Stiff
Color-codedNo
Handle MaterialStainless Steel Wire
Handle TypeTwisted Wire
Tip TypeFan Tip
ScentUnscented
SterilityNon-sterile
ColorWhite
Diameter (English) Bristle0.5 in.
Diameter (Metric) Bristle12 mm
For Use With (Application)For cleaning test tubes
Length (English)9 in.
Length (English) Brush3 in.
Length (Metric)230 mm
Length (Metric) Brush80 mm
Quantity1 Ea.
Weight (Metric)3 g
Unit SizeEA