Porcelain Buchner Funnels with Fixed Perforated Plates
Porcelain Buchner Funnels with Fixed Perforated Plates
Actual product may vary
Fisherbrand
Porcelain Buchner Funnels with Fixed Perforated Plates
For suction filtration applications
Have Questions?
Catalog NumberCapacity (Metric)Diameter (Metric) DiscDiameter (Metric) Plate
FISFB966F320 mL90mm100mm
Catalog number FISFB966F
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
320 mL
Diameter (Metric) Disc:
90mm
Diameter (Metric) Plate:
100mm
For suction filtration applications
  • Autoclavable
  • Highly resistant to chemical attack
  • Without cover
  • Glazed inside and outside with the exception of the rim
Specifications
Diameter (Metric)Top10.5cm
Capacity (Metric)320 mL
MaterialPorcelain
Height (Metric)16 cm
AutoclavableAutoclavable
Diameter (Metric) Plate100mm
For Use With (Application)Suction filtration applications
Diameter (Metric) Disc90mm
IncludesFixed perforated plate
QuantityEach
Wall Thickness4.5mm
Weight (Metric)0.41kg
Product TypeBuchner Funnel
Unit Size