Porcelain Buchner Funnels with Fixed Perforated Plates
Porcelain Buchner Funnels with Fixed Perforated Plates
Actual product may vary
Fisher Catalogue
Porcelain Buchner Funnels with Fixed Perforated Plates
For suction filtration applications
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Diameter (Metric) DiscDiameter (Metric) Plate
FISFB966B30 mL35 - 40mm41 mm
FISFB966F320 mL90mm100mm
FISFB966G550 mL110mm114mm
FISFB966H700 mL110 - 124mm126mm
FISFB966J1060 mL142 - 150mm151mm
FISFB966K1860 mL185mm186mm
Catalog number FISFB966B
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
30 mL
Diameter (Metric) Disc:
35 - 40mm
Diameter (Metric) Plate:
41 mm
For suction filtration applications
  • Autoclavable
  • Highly resistant to chemical attack
  • Without cover
  • Glazed inside and outside with the exception of the rim
Specifications
Diameter (Metric)Top5.2cm
Capacity (Metric)30 mL
MaterialPorcelain
Height (Metric)8.9 cm
AutoclavableAutoclavable
Diameter (Metric) Plate41 mm
For Use With (Application)Suction filtration applications
Diameter (Metric) Disc35 - 40mm
IncludesFixed perforated plate
QuantityEach
Wall Thickness3.5mm
Weight (Metric)0.07kg
Product TypeBuchner Funnel
Unit Size