Porcelain Pestles
Porcelain Pestles
Actual product may vary
Fisher Catalogue
Porcelain Pestles
For hand comfort in grinding
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)Length (English)Length (Metric)
FISFB961M150mL Mortar6.2 in.15.7cm
FISFB961O400mL Mortar7.6 in.19.4cm
Catalog number FISFB961M
View Price:Sign in to see your account pricing. Need an account? Register with us today.
For Use With (Equipment):
150mL Mortar
Length (English):
6.2 in.
Length (Metric):
15.7cm
For hand comfort in grinding
  • Unglazed on grinding surface
  • Autoclavable
  • Oversized
Specifications
FinishUnglazed Grinding Surface
AutoclavableAutoclavable
Product TypePestle
MaterialPorcelain
For Use With (Equipment)150mL Mortar
For Use With (Application)For hand comfort in grinding
Length (Metric)15.7cm
Length (English)6.2 in.
Unit Size