Reusable Glass Narrow-Mouth Erlenmeyer Flasks
Reusable Glass Narrow-Mouth Erlenmeyer Flasks
Actual product may vary
Fisherbrand
Reusable Glass Narrow-Mouth Erlenmeyer Flasks
Long, tapered outside contour minimizes chipping
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
FISFB50010001000 mL
FISFB500125125 mL
FKS154461431000 mL
FISFB500500500 mL
FKS15436143500 mL
FISFB500250250 mL
Catalog number FISFB5001000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
1000 mL
Fisherbrand™ Reusable Glass Narrow-Mouth Erlenmeyer Flasks are useful for measuring and mixing solutions where a high degree of accuracy is not necessary. Long, tapered outside contour minimizes chipping.
  • Reinforced top tooled with a rounded finish
  • Thick-walled body
  • White ceramic enamel marking spot
Specifications
Closure MaterialRubber
DescriptionTop style: Heavy duty tooled with capacity scale
Diameter (English) Outer5 in.
Capacity (Metric)1000 mL
MaterialBorosilicate Glass
Graduation Range250 to 1000 mL
Graduations50 mL
Height (English)8.38 in.
Height (Metric)21.3 cm
No. of Positions1
Writing SurfaceMatte Finish Ceramic Enamel Marking
DisposableReusable
Mouth StyleNarrow
Diameter (Metric) Outer128 mm
Closure Size9
Unit Size