Search Thermo Fisher Scientific
Catalog Number | Diameter (Metric) | Height (Metric) | Shape |
---|---|---|---|
FISFB0875713A | 60 mm | 15 mm | Round |
FISFB0875714 | 150 mm | 15 mm | Round |
FISFB0875712 | 100 mm | 15 mm | Round |
Note: Cat. No. FB0875713 has an ISO mark for use in automation.