Standard Class B Graduated Cylinders
Standard Class B Graduated Cylinders
Actual product may vary
Fisher Catalogue
Standard Class B Graduated Cylinders
Calibrated To Deliver
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)GraduationsTolerance MetricSubdivision
FIS08-550E100 mL1 mL±1 mL1mL
FIS08-550B10 mL0.2 mL±0.2 mL0.2mL
FIS08-550D50 mL1 mL±0.5 mL1mL
FIS08-550A5 mL0.1 mL±0.1 mL0.1mL
FIS08-550C25 mL0.5 mL±0.34 mL0.5mL
FIS08-550G500 mL5 mL±4 mL5mL
Catalog number FIS08-550E
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
100 mL
Graduations:
1 mL
Tolerance Metric:
±1 mL
Subdivision:
1mL
Calibrated To Deliver
  • White-ceramic graduations
  • Heavy round bases
  • Handy pouring spouts
  • Accuracy is assured even at the lowest graduations; first 10% volume is not graduated
Specifications
Base TypeRound
CalibrationTo Deliver
Scale ColorWhite
ShapeRound
ClassB
Height (English)10.2 in.
Height (Metric)260 mm
Tolerance Metric±1 mL
TypeGraduated Cylinder
MaterialBorosilicate Glass
Capacity (Metric)100 mL
For Use With (Application)Accuracy assured even at lowest graduations
ScaleWhite ceramic graduations
Graduations1 mL
Subdivision1mL
Unit Size