Check Order Status
Fused-Quartz Crucibles
Actual product may vary
Fisher Catalogue
Fused-Quartz Crucibles
Withstand temperatures up to 1250°C (2282°F)
Have Questions?
Change view
Catalog Number
Capacity (Metric)
Diameter (Metric) Outer
Height (Metric)
FIS08-072D
30 mL
4.3cm
3.8cm
FIS08-072F
100 mL
6.2cm
6.6cm
2 Options
Catalog number
FIS08-072D
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
30 mL
Diameter (Metric) Outer:
4.3cm
Height (Metric):
3.8cm
Withstand temperatures up to 1250°C (2282°F)
Withstand severe temperature changes
Insoluble in water and acids except hydrofluoric and phosphoric acids
Covers sold separately
Specifications
Color
Clear
Description
High purity fused quartz
Material
Quartz
Max. Temperature (Metric)
1250°C
Thickness (Metric)
2mm
Capacity (Metric)
30 mL
Diameter (Metric) Outer
4.3cm
Height (Metric)
3.8cm
Product Type
Crucible
Unit Size