Rubber Vacuum Hose
Rubber Vacuum Hose
Actual product may vary
Rubber Vacuum Hose
Thick-walled red rubber vacuum hose. Welch Ilmvac™ Rubber Vacuum Hose well suited for laboratory vacuum pump processes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTubing Size
FIL828310-48mm ID, 18mm OD
WEL828310-48mm ID, 18mm OD
Catalog number FIL828310-4
Tubing Size:
8mm ID, 18mm OD
  • Economical
  • Flexible
  • Resilient
  • Thick-walled
Specifications
Diameter (Metric) Inner8 mm
MaterialRubber
Diameter (Metric) Outer18 mm
Overall Length (Metric)100 cm
Packaging Quantity1 Each
Tubing Size8mm ID, 18mm OD
ColorRed
Product TypeVacuum Hose
Unit Size