Check Order Status
Gentamycin Sulfate (White Powder), Fisher BioReagents
Element Search
Structure Search
Fisher Chemical
Gentamycin Sulfate (White Powder), Fisher BioReagents
For selecting transformed cells containing gentamycin resistance gene
Have Questions?
Catalog Number
Quantity
FGCBP918-1
1 g
Catalog number
FGCBP918-1
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS
7697-37-2
IUPAC Name
nitric acid
Molecular Formula
HNO3
InChI Key
GRYLNZFGIOXLOG-UHFFFAOYSA-N
SMILES
O[N+]([O-])=O
View more
Specifications
Specification Sheet
Chloride (Cl)
=<0.5 ppm
Copper (Cu)
=<0.05 ppm
Cadmium (Cd)
=<0.01 ppm
Arsenic (As)
=<0.01 ppm
Appearance (Form)
Liquid
View more
Gentamycin Sulfate inhibits protein synthesis, preventing microbial contamination in tissue culture applications. It is also used to confer antibiotic resistance/sensitivity in molecular biology procedures.