HEPES (Fine White Crystals/Molecular Biology), Fisher BioReagents™
HEPES (Fine White Crystals/Molecular Biology), Fisher BioReagents™
HEPES (Fine White Crystals/Molecular Biology), Fisher BioReagents™
HEPES (Fine White Crystals/Molecular Biology), Fisher BioReagents™
Fisher Chemical
HEPES (Fine White Crystals/Molecular Biology), Fisher BioReagents™
Commonly used buffering agent.
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
1 kg
5 kg
Packaging:
View all
Poly Bottle
Catalog number FGCBP310-500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Poly Bottle
Request bulk or custom format
Chemical Identifiers
CAS4101-30-8
IUPAC Name1-(2-amino-4,5-dimethoxyphenyl)ethan-1-one
Molecular FormulaC10H13NO3
InChI KeyKGKWXEGYKGTMAK-UHFFFAOYSA-N
SMILESCOC1=CC(N)=C(C=C1OC)C(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)103.0-109.0?C
FormPowder
Assay (GC)≥97.5%
Appearance (Color)Yellow
Commonly used buffering agent
HEPES is commonly used as a buffering agent.