Check Order Status
Sodium Dodecyl Sulfate (SDS), White Powder, Electrophoresis, Fisher BioReagents™
Element Search
Structure Search
Fisher Chemical
Sodium Dodecyl Sulfate (SDS), White Powder, Electrophoresis, Fisher BioReagents™
For protein extraction procedures and SDS polyacrylamide gels
Have Questions?
Change view
Quantity:
500 g
100 g
Packaging:
View all
Poly Bottle
Catalog number
FGCBP166-500
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Poly Bottle
Request bulk or custom format
Chemical Identifiers
CAS
9003-01-4
IUPAC Name
Poly(acrylic acid)
Molecular Formula
(C3H4O2)n
SMILES
OC(=O)C(-*)C-*
MDL Number
MFCD00084394
Specifications
Specification Sheet
Infrared spectrum
Conforms
Heavy metals
=<10 ppm (Hg, Pb, As, Sb)
Viscosity
(neutralized soln., 0.5% dispersion)
Appearance (Color)
White
Loss on drying
=<2 %
View more
For protein extraction procedures and SDS polyacrylamide gels
DNase and RNase-free detergent for protein extraction procedures and polyacrylamide gel electrophoresis
SDS is a commonly used detergent in protein purification and polyacrylamide gel electrophoresis (PAGE)
SDS denatures proteins by disrupting the non-covalent bonds of the protein
Functionally tested for use in electrophoresis
Intended for research use only; not for human or animal diagnostic purposes