Triton™ X-100 (Electrophoresis), Fisher BioReagents™
Triton™ X-100 (Electrophoresis), Fisher BioReagents™
Triton™ X-100 (Electrophoresis), Fisher BioReagents™
Triton™ X-100 (Electrophoresis), Fisher BioReagents™
Fisher Chemical
Triton™ X-100 (Electrophoresis), Fisher BioReagents™
For solubilization and permeabilization of membranes
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
Packaging:
HDPE Bottle
Catalog number FGCBP151-500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Packaging:
HDPE Bottle
Request bulk or custom format
Chemical Identifiers
CAS9005-84-9
IUPAC Name(2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-{[(2R,3S,4R,5R,6S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}oxane-3,4,5-triol
Molecular FormulaC12H22O11
InChI KeyGUBGYTABKSRVRQ-ASMJPISFSA-N
SMILESOC[C@H]1O[C@H](O[C@H]2[C@H](O)[C@@H](O)[C@@H](O)O[C@@H]2CO)[C@H](O)[C@@H](O)[C@@H]1O
View more
SpecificationsSpecification SheetSpecification Sheet
SensitivityPasses test
Appearance (Form)Clear to slightly hazy solution
This nonionic detergent is used to solubilize membranes, permeabilize unfixed eukaryotic membranes, and is a component of lysis buffers for DNA extraction.