Check Order Status
Methylene Blue, Fisher BioReagents™
Element Search
Structure Search
Methylene Blue, Fisher BioReagents™
Have Questions?
Catalog Number
Quantity
FGCBP117-100
100 g
Catalog number
FGCBP117-100
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or Material
Methylene Blue
Melting Point
190°C
CAS
7220-79-3
ChemAlert Storage Symbol
Gray
Health Hazard 2
WARNING!
View more