Check Order Status
Sodium Tetraphenylboron, 0.02 M, Ricca Chemical
Element Search
Structure Search
Sodium Tetraphenylboron, 0.02 M, Ricca Chemical
Have Questions?
Catalog Number
Quantity
FCS7855-32
1 L
Catalog number
FCS7855-32
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Request bulk or custom format
Specifications
Chemical Name or Material
Sodium Tetraphenylboron
Color
Colorless
CAS
143-66-8
CAS Min %
97.0
CAS Max %
100.0
View more