Check Order Status
Platinum (IV) Chloride Solution (10% w/v), Fisher Chemical
Element Search
Structure Search
Platinum (IV) Chloride Solution (10% w/v), Fisher Chemical
Have Questions?
Catalog Number
Quantity
FCMSP119-25
25 mL
Catalog number
FCMSP119-25
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 mL
Request bulk or custom format
Specifications
Chemical Name or Material
Platinum (IV) Chloride Solution
Color
Yellow to Orange
Name Note
10% w/v
CAS
16941-12-1
ChemAlert Storage Symbol