Check Order Status
Lewatit NM 60 Ion Exchange Resin (Analytical Grade/Certified), Fisher Chemical
Element Search
Structure Search
Fisher Chemical
Lewatit NM 60 Ion Exchange Resin (Analytical Grade/Certified), Fisher Chemical
Have Questions?
Catalog Number
Quantity
Packaging
FCMR276-500
500 g
Poly Bottle
Catalog number
FCMR276-500
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Poly Bottle
Request bulk or custom format
Specifications
Chemical Name or Material
Lewatit NM 60 ion exchange resin
Physical Form
Solid
Name Note
Analytical Grade
CAS
69011-49-0
ChemAlert Storage Symbol
Gray
View more