Check Order Status
Ferric Chloride Hexahydrate (Lumps/Certified ACS), Fisher Chemical™
Element Search
Structure Search
Fisher Chemical
Ferric Chloride Hexahydrate (Lumps/Certified ACS), Fisher Chemical™
Have Questions?
Catalog Number
Quantity
Packaging
FCMI88-100
100 g
Poly Bottle
Catalog number
FCMI88-100
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Poly Bottle
Request bulk or custom format
Specifications
Boiling Point
280°C
Chemical Name or Material
Ferric Chloride Hexahydrate
Melting Point
37°C
CAS
10025-77-1
ChemAlert Storage Symbol