Check Order Status
DMAC, Dimethylacetamide, GC Headspace Grade, Thermo Scientific™
Element Search
Structure Search
Fisher Chemical
DMAC, Dimethylacetamide, GC Headspace Grade, Thermo Scientific™
DMAC processed to high purity for accurate and repeatable determination of trace levels of residual solvents.
Have Questions?
Catalog Number
Quantity
Packaging
FCMD160-1
1 L
Glass Bottle
Catalog number
FCMD160-1
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Specifications
Chemical Name or Material
DMAC, Dimethylacetamide
Absorbance
0.300 AU Maximum (275nm), 0.080 AU Maximum (300nm), 0.005 AU Maximum (350nm)
CAS
127-19-5
Color
Colorless
Recommended Storage
Room temperature, packaged under inert atmosphere
View more
Every lot is tested to ensure interference free baselines for interference free determination of Class 1, 2, and 3 residual solvents
1 ppm max. of Class 1 Solvents, 10 ppm max. of Class 2 solvents, and 50 ppm max of Class 3 residual solvents
This product is UV Absorbance Tested to ensure low organic contamination
Processed for low water content to facilitate the extraction of organic volatile impurities, or VOC