Dextrose (D-Glucose), Anhydrous (Granular Powder/Certified ACS), Fisher Chemical™
Dextrose (D-Glucose), Anhydrous (Granular Powder/Certified ACS), Fisher Chemical™
Dextrose (D-Glucose), Anhydrous (Granular Powder/Certified ACS), Fisher Chemical™
Dextrose (D-Glucose), Anhydrous (Granular Powder/Certified ACS), Fisher Chemical™
Dextrose (D-Glucose), Anhydrous (Granular Powder/Certified ACS), Fisher Chemical™
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
Packaging:
Poly Bottle
Catalog number FCMD16-1
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 kg
Packaging:
Poly Bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialDextrose (D-Glucose) Anhydrous
CAS50-99-7
ChemAlert Storage SymbolGray
Health Hazard 2CAUTION!
Health Hazard 3Emergency Overview
May cause skin, eye, and respiratory tract irritation. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. Obtain medical attention. .
NFPA
Health:1
Flammability:0
Instability:0
View more