Check Order Status
Ammonium Hydroxide (Certified ACS Plus), Fisher Chemical™
Element Search
Structure Search
Fisher Chemical
Ammonium Hydroxide (Certified ACS Plus), Fisher Chemical™
Have Questions?
Change view
Quantity:
6 x 2.5 L
2.5 L
Packaging:
Glass Bottle
Catalog number
FCMA669C-212
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
6 x 2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Specifications
Chemical Name or Material
Ammonium Hydroxide
pH
12
Cleanup Symbol
Caustic
Color
Colorless
Name Note
about 14.8N
View more