Check Order Status
Ammonium Formate (Crystalline/Certified ACS), Fisher Chemical
Element Search
Structure Search
Fisher Chemical
Ammonium Formate (Crystalline/Certified ACS), Fisher Chemical
Have Questions?
Catalog Number
Quantity
Packaging
FCMA666-500
500 g
Poly Bottle
Catalog number
FCMA666-500
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Poly Bottle
Request bulk or custom format
Specifications
Boiling Point
180°C
Chemical Name or Material
Ammonium Formate
Melting Point
115°C
CAS
540-69-2
ChemAlert Storage Symbol
Gray
View more