Check Order Status
Nitric Acid (TraceMetal™ Grade), Fisher Chemical™
Element Search
Structure Search
Fisher Chemical
Nitric Acid (TraceMetal™ Grade), Fisher Chemical™
Have Questions?
Change view
Quantity:
500 mL
Packaging:
HDPE Bottle
Catalog number
FCMA509-P500
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Packaging:
HDPE Bottle
Request bulk or custom format
Specifications
Chemical Name or Material
Nitric Acid
Shelf Life
2 years
pH
1.0
Cleanup Symbol
Acid
Color
Colorless Yellow
View more
Nitric Acid (TraceMetal™ Grade)
Ideal for analysis by IC, GF or Flame AA, ICP-OES and ICP-MS.
Evaluated for 65 metals at < 1 ppb concentrations.
Manufactured,purified, and packaged in a Class 1000 clean room.
Packaged in non-breakable polypropylene bottles to maintain product integrity and to eliminate metal leaching from glass bottles.
Two year shelf life from time of manufacture - available on COA. 45 mm bottle neck size