Check Order Status
Hydrochloric Acid, Certified ACS Plus, 36.5 to 38.0%, Fisher Chemical™
Element Search
Structure Search
Fisher Chemical
Hydrochloric Acid, Certified ACS Plus, 36.5 to 38.0%, Fisher Chemical™
Have Questions?
Change view
Quantity:
2.5 L
500 mL
Packaging:
Glass Bottle
Catalog number
FCMA144-212
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Specifications
pH
<1
Cleanup Symbol
Acid
CAS
7647-01-0
Concentration
36.5 to 38.0% (w/w)
Density
1.27 (Air=1.0)
View more