Check Order Status
Gentamycin Sulfate (White Powder), Fisher BioReagents
Element Search
Structure Search
Fisher Chemical
Gentamycin Sulfate (White Powder), Fisher BioReagents
For selecting transformed cells containing gentamycin resistance gene
Have Questions?
Catalog Number
Quantity
FBRBP918-1
1 g
Catalog number
FBRBP918-1
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS
1532-71-4
IUPAC Name
1-bromoisoquinoline
Molecular Formula
C9H6BrN
InChI Key
YWWZASFPWWPUBN-UHFFFAOYSA-N
SMILES
BrC1=C2C=CC=CC2=CC=N1
View more
Specifications
Specification Sheet
Melting point
42°C to 48°C
Appearance (Color)
Yellow to brown
Appearance (Form)
Powder and/or chunks
Infrared spectrum
Conforms
HPLC
>=95.0 %
Gentamycin Sulfate inhibits protein synthesis, preventing microbial contamination in tissue culture applications. It is also used to confer antibiotic resistance/sensitivity in molecular biology procedures.