Check Order Status
Sodium Acetate Anhydrous (White Crystals or Granular Powder), Fisher BioReagents
Element Search
Structure Search
Fisher Chemical
Sodium Acetate Anhydrous (White Crystals or Granular Powder), Fisher BioReagents
Have Questions?
Change view
Quantity:
500 g
1 kg
Packaging:
View all
Poly Bottle
Catalog number
FBRBP333-500
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Poly Bottle
Request bulk or custom format
Specifications
Chemical Name or Material
Sodium Acetate Anhydrous
Melting Point
324°C
CAS
127-09-3
ChemAlert Storage Symbol
Gray
Color
White
View more
Sodium Acetate is added to DNA reactions to facilitate ethanol precipitation of the DNA.