Check Order Status
Sodium Bicarbonate (Fine White Powder), Fisher BioReagents™
Element Search
Structure Search
Fisher Chemical
Sodium Bicarbonate (Fine White Powder), Fisher BioReagents™
CAS: 144-55-8 | CHNaO3 | 84.01 g/mol
Have Questions?
Change view
Quantity:
500 g
1 kg
Packaging:
Poly Bottle
Catalog number
FBRBP328-500
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Poly Bottle
Request bulk or custom format
Chemical Identifiers
CAS
3188-00-9
IUPAC Name
2-methyloxolan-3-one
Molecular Formula
C5H8O2
InChI Key
FCWYQRVIQDNGBI-UHFFFAOYNA-N
SMILES
CC1OCCC1=O
View more
Specifications
Specification Sheet
Form
Liquid
Identification (FTIR)
Conforms
Assay (GC)
≥98.0%
Refractive Index
1.4270-1.4310 @ 20?C
Appearance (Color)
Clear colorless to pale yellow
Sodium Bicarbonate (Fine White Powder)
This sodium salt is used in buffers for molecular biology and cell culture applications.